
1-Dodecyl-3-methylimidazolium bis(trifluoromethanesulphonyl)imide ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:404001-48-5
:
Formula:C₁₈H₃₁F₆N₃O₄S₂
MDL Number:MFCD12022372
:531.57 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Brown clear liquid
- Purity (HPLC):min. 93.0 area%
- Purity (Ion exchange titration):min. 95.0 %
Specification Test Results
Frequently Bought Together