


1,5-Diaminonaphthalene ≥99.5% (by GC), purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:2243-62-1
:
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004029
:158.2 g/mol
EINECS:218-817-8
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 99.5 %
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:189.0 to 194.0 deg-C
Specification Test Results
Frequently Bought Together