3,5-Dibromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:942132-66-3
:
Formula:C₂₄H₁₆Br₂
MDL Number:MFCD32263474
:464.20 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:145.0 to 149.0 deg-C
Specification Test Results
Frequently Bought Together