
2,5-Dimethoxyphenol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:18113-18-3
:
Formula:C₈H₁₀O₃
MDL Number:MFCD00094237
:154.1600 g/mol
Boiling Pt:135°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together