Order Entry
Italy
ContactUsLinkComponent
Dicyclohexyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by GC)
Irritant
Dicyclohexyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by GC)
Catalog # TCIAD5664-250MG
Supplier:  TCI
CAS Number:  
Irritant
Dicyclohexyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by GC)
Catalog # TCIAD5664-250MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    193.0 to 197.0 deg-C

Specification Test Results

About this item

 :
 :536.78 g/mol
 :GHS07
CAS Number:1070663-78-3
Formula:C₃₅H₅₃O₂P
MDL Number:MFCD11973797