Order Entry
Italy
ContactUsLinkComponent
Di-tert-butyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by HPLC)
Irritant
Di-tert-butyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by HPLC)
Catalog # TCIAD5663-250MG
Supplier:  TCI
CAS Number:  
Irritant
Di-tert-butyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by HPLC)
Catalog # TCIAD5663-250MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    169.0 to 173.0 deg-C

Specification Test Results

About this item

 :
 :484.70 g/mol
 :GHS07
CAS Number:1160861-53-9
Formula:C₃₁H₄₉O₂P
MDL Number:MFCD13181930