5,7-Di-tert-butyl-3-(3,4-dimethylphenyl)benzofuran-2(3H)-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:164391-52-0
:
Formula:C₂₄H₃₀O₂
MDL Number:MFCD30729989
:350.50 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:131.0 to 135.0 deg-C
Specification Test Results
Frequently Bought Together