Order Entry
Italy
ContactUsLinkComponent
 
 
4,4'-Dibromotriphenylamine ≥98.0% (by GC, titration analysis), purified by sublimation
 :  TCI
 
 
4,4'-Dibromotriphenylamine ≥98.0% (by GC, titration analysis), purified by sublimation
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

 

Frequently Bought Together