Order Entry
Italy
ContactUsLinkComponent
2,4-Diphenyl-6-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine ≥98.0% (by GC)
2,4-Diphenyl-6-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine ≥98.0% (by GC)
Supplier:  TCI
Certificates
2,4-Diphenyl-6-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine ≥98.0% (by GC)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    202.0 to 206.0 deg-C

Specification Test Results

Frequently Bought Together