Order Entry
Italy
ContactUsLinkComponent
 
Doxylamine succinate ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
Doxylamine succinate ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 % (total of Ion-Pair)
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    109.0 to 113.0 deg-C

 

Frequently Bought Together