Order Entry
Italy
ContactUsLinkComponent
 
Irritant
Diglycidyl 4-Cyclohexene-1,2-dicarboxylate
Catalog # TCIAD5576-25G
Supplier:  TCI
CAS Number:  
Irritant
Diglycidyl 4-Cyclohexene-1,2-dicarboxylate
Catalog # TCIAD5576-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Oxirane oxygen:
    9.0 to 12.0 %

Specification Test Results

About this item

 :
 :282.29 g/mol
EINECS:244-435-6
 :GHS07
Storage Temperature:Refrigerator
CAS Number:21544-03-6
Formula:C₁₄H₁₈O₆
MDL Number:MFCD00209575