
3,3-Difluoropiperidine hydrochloride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:496807-97-7
:
Formula:C5H10ClF2N
MDL Number:MFCD03452800
:157.59 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together