


1,3-Dioxolane ≥98.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:646-06-0
:
Formula:C₃H₆O₂
MDL Number:MFCD00003207
:74.08 g/mol
Boiling Pt:74°C
Flash Pt:–4°C
EINECS:211-463-5
UN:1166
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together