Inserisci ordine
Italy
ContactUsLinkComponent
2,2-Difluoropropionic acid ≥98.0% (by GC, titration analysis)
Corrosive
2,2-Difluoropropionic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD5531-1G
Supplier:  TCI
CAS Number:  
Corrosive
2,2-Difluoropropionic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD5531-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Conf:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    43.0 to 47.0 deg-C

Specification Test Results

About this item

 :
 :110.06 g/mol
UN:3261
ADR:8,II
 :GHS05
CAS Number:373-96-6
Formula:C₃H₄F₂O₂
MDL Number:MFCD03093764