
4-(4,6-Dimorpholino-1,3,5-triazin-2-yl)aniline ≥98.0% (by titrimetric analysis)
: TCI
:1197159-91-3
:
:C₁₇H₂₂N₆O₂
:MFCD27921193
:342.40 g/mol
:GHS07
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together