
1,3-Di-tert-butylimidazolium hydrogencarbonate ≥98.0% (by HPLC) (contains varying amounts of 1,3-Di-tert-butylimidazolium-2-carboxylate)
Catalog # TCIAD5513-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
Specification Test Results
About this item
:
:242.32 g/mol
:GHS07
CAS Number:479641-76-4
Formula:C₁₂H₂₂N₂O₃