Order Entry
Italy
ContactUsLinkComponent
(R)-1,4-Dioxaspiro[4.5]decane-2-carboxaldehyde ≥98.0% (by GC)
(R)-1,4-Dioxaspiro[4.5]decane-2-carboxaldehyde ≥98.0% (by GC)
Catalog # TCIAD5511-5G
Supplier:  TCI
CAS Number:  
(R)-1,4-Dioxaspiro[4.5]decane-2-carboxaldehyde ≥98.0% (by GC)
Catalog # TCIAD5511-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +73.0 to +81.0 deg (C=1, CHCl3)
  • Acid Value:
    max. 5.0

Specification Test Results

About this item

 :
 :170.21 g/mol
Boiling Pt:94°C
CAS Number:78008-36-3
Formula:C₉H₁₄O₃
MDL Number:MFCD03093997