5,10-Dibromonaphtho[1,2-c:5,6-c']bis([1,2,5]oxadiazole) ≥98.0% (by HPLC)
Catalog # TCIAD5496-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):14.30 to 15.80 %
Specification Test Results
About this item
:
:369.96 g/mol
CAS Number:1437229-17-8
Formula:C₁₀H₂Br₂N₄O₂