Order Entry
Italy
ContactUsLinkComponent
 
2-Decyl-7-phenyl[1]benzothieno[3,2-b][1]benzothiophene ≥99.5% (by HPLC)
 :  TCI
 
2-Decyl-7-phenyl[1]benzothieno[3,2-b][1]benzothiophene ≥99.5% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 99.5 area%
  • Hole Mobility (mu FET):
    min. 10.0 cm2/Vs (ODTS Si/SiO2 substrate)

 

Frequently Bought Together