3,3'-Di(dibenzothiophen-4-yl)-1,1'-biphenyl ≥98.0% (by HPLC)
TCIAD5473-1G
: TCI
:
- Pk:1 g/mol
- Seq:9220
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Carbon):82.00 to 85.00 %
:
:518.69 g/mol
:1128045-14-6
:C₃₆H₂₂S₂
:MFCD32062886