Order Entry
Italy
ContactUsLinkComponent
N,N'-Di(1-naphthyl)-N,N',9,9-tetraphenyl-9H-fluorene-2,7-diamine ≥98.0% (by HPLC)
N,N'-Di(1-naphthyl)-N,N',9,9-tetraphenyl-9H-fluorene-2,7-diamine ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
N,N'-Di(1-naphthyl)-N,N',9,9-tetraphenyl-9H-fluorene-2,7-diamine ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    237.0 to 241.0 deg-C

Specification Test Results

Frequently Bought Together