

(3,3'-Difluoro-[2,2'-bithiophene]-5,5'-diyl)bis(trimethylstannane) ≥98.0% (by HPLC)
TCIAD5436-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
:
:527.85 g/mol
:3146
:6.1,I
:GHS06
:GHS09
:1619967-09-7
:C₁₄H₂₀F₂S₂Sn₂
:MFCD30543603