
2,7-Dimethoxy-9H-carbazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:61822-18-2
:
Formula:C₁₄H₁₃NO₂
MDL Number:MFCD28154716
:227.26 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together