Order Entry
Italy
ContactUsLinkComponent
 
 
4,5-Dichloro-2-methyl-2H-pyridazin-3-one ≥98.0% (by GC)
 :  TCI
 
 
4,5-Dichloro-2-methyl-2H-pyridazin-3-one ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    87.0 to 91.0 deg-C

 

Frequently Bought Together