4,4'-Di-tert-butylbenzophenone ≥95.0% (by GC)
TCIAD5308-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 95.0 %
- Melting point:132.0 to 136.0 deg-C
:
:294.4306 g/mol
:158°C
:15796-82-4
:C₂₁H₂₆O
:MFCD01319937