

2,2-Difluoroethyl trifluoromethanesulphonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:74427-22-8
:
Formula:C₃H₃F₅O₃S
MDL Number:MFCD02093342
:214.11 g/mol
Boiling Pt:118°C
UN:2920
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together