
5,7-dihydroindolo[2,3-b]carbazole ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD5286-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
Melting Pt:360°C
:256.308 g/mol
:GHS07
CAS Number:111296-90-3
Formula:C₁₈H₁₂N₂
MDL Number:MFCD10687177