2,4-Diphenyl-6-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 2-[4-(4,6-Diphenyl-1,3,5-triazin-2-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,6-Diphenyl-1,3,5-triazine-2-yl)phenylboronic acid Pinacol ester, 2,4-Diphenyl-6-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1,3,5-triazine
CAS Number:1219956-23-6
:
Formula:C₂₇H₂₆BN₃O₂
MDL Number:MFCD29918901
:435.33 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:210.0 to 214.0 deg-C
Specification Test Results
Frequently Bought Together