
2,7-Dibromo-9-(4-bromophenyl)-9H-carbazole ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1313900-20-7
:
Formula:C₁₈H₁₀Br₃N
MDL Number:MFCD30469057
:480.00 g/mol
Melting Pt:136°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together