Order Entry
Italy
ContactUsLinkComponent
1,5-Dibromo-2,4-difluorobenzene ≥98.0% (by GC)
Irritant
1,5-Dibromo-2,4-difluorobenzene ≥98.0% (by GC)
Catalog # TCIAD5259-1G
Supplier:  TCI
CAS Number:  
Irritant
1,5-Dibromo-2,4-difluorobenzene ≥98.0% (by GC)
Catalog # TCIAD5259-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    29.0 to 33.0 deg-C

Specification Test Results

About this item

 :
Boiling Pt:102°C
 :271.8849 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:28342-75-8
Formula:C₆H₂Br₂F₂
MDL Number:MFCD08458122