Order Entry
Italy
ContactUsLinkComponent
Di(thiophen-2-yl)methane ≥90.0% (by GC)
Di(thiophen-2-yl)methane ≥90.0% (by GC)
Catalog # TCIAD5228-500MG
Supplier:  TCI
CAS Number:  
Di(thiophen-2-yl)methane ≥90.0% (by GC)
Catalog # TCIAD5228-500MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 90.0 %
  • Melting point:
    44.0 to 48.0 deg-C

Specification Test Results

About this item

 :
 :180.28 g/mol
Boiling Pt:267°C
CAS Number:4341-34-8
Formula:C₉H₈S₂
MDL Number:MFCD00041212