Order Entry
Italy
ContactUsLinkComponent
 
3,7-Dibromo-10-hexylphenothiazine ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
3,7-Dibromo-10-hexylphenothiazine ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    54.0 to 58.0 deg-C

 

Frequently Bought Together