4,4'-Dibromobenzophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3988-03-2
:
Formula:C₁₃H₈Br₂O
MDL Number:MFCD00016330
:340.01 g/mol
Boiling Pt:395°C
EINECS:223-632-0
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:174.0 to 178.0 deg-C
Specification Test Results
Frequently Bought Together