Order Entry
Italy
ContactUsLinkComponent
Diethyl (4-nitrobenzyl)phosphonate ≥97.0% (by GC)
Diethyl (4-nitrobenzyl)phosphonate ≥97.0% (by GC)
Catalog # TCIAD5208-5G
Supplier:  TCI
CAS Number:  
undefined
Diethyl (4-nitrobenzyl)phosphonate ≥97.0% (by GC)
Catalog # TCIAD5208-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :273.225 g/mol
Boiling Pt:153°C
CAS Number:2609-49-6
Formula:C₁₁H₁₆NO₅P
MDL Number:MFCD00047747