Order Entry
Italy
ContactUsLinkComponent
1,1-Dichlorosilacyclobutane ≥98.0% (by GC, titration analysis)
Flammable
Corrosive
1,1-Dichlorosilacyclobutane ≥98.0% (by GC, titration analysis)
Catalog # TCIAD5198-1G
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
1,1-Dichlorosilacyclobutane ≥98.0% (by GC, titration analysis)
Catalog # TCIAD5198-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :141.07 g/mol
Boiling Pt:115°C
Flash Pt:20°C
EINECS:219-084-7
UN:2988
ADR:4.3,I
 :GHS02
 :GHS05
CAS Number:2351-33-9
Formula:C₃H₆Cl₂Si
MDL Number:MFCD00053537