
3,6-Dihydroxyflavone ≥98.0% (by HPLC)
Catalog # TCIAD5191-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:238.0 to 242.0 deg-C
Specification Test Results
About this item
:
:254.24 g/mol
:GHS07
CAS Number:108238-41-1
Formula:C₁₅H₁₀O₄
MDL Number:MFCD00143082