1H-Pyrrolo[3,2-d]pyrimidine-2,4(3H,5H)-dione ≥98.0% (by HPLC)
Catalog # TCIAD5183-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
Specification Test Results
About this item
:
:151.12 g/mol
CAS Number:65996-50-1
Formula:C₆H₅N₃O₂
MDL Number:MFCD08059275