
1,7-Dichloro-4-methoxyisoquinoline ≥98.0% (by GC)
Catalog # TCIAD5182-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:137.0 to 142.0 deg-C
Specification Test Results
About this item
:
:228.07 g/mol
:GHS07
CAS Number:630423-36-8
Formula:C₁₀H₇Cl₂NO
MDL Number:MFCD17012717