
4,5-Dibromo-3(2H)-pyridazinone ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5788-58-9
:
Formula:C₄H₂Br₂N₂O
MDL Number:MFCD00023641
:253.89 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together