
4,5-Difluoro-2-methoxybenzonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:425702-28-9
:
Formula:C₈H₅F₂NO
MDL Number:MFCD06660346
:169.13 g/mol
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:97.0 to 101.0 deg-C
Specification Test Results
Frequently Bought Together