4,4'-(2,3-Dihydrothieno[3,4-b][1,4]dioxine-5,7-diyl)bis[N,N-bis(4-methoxyphenyl)aniline] ≥98.0% (by total nitrogen basis)
Catalog # TCIAD5155-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:748.89 g/mol
CAS Number:1622008-73-4
Formula:C₄₆H₄₀N₂O₆S
MDL Number:MFCD28369275