
5,6-Dichloro-1H-1,3-benzimidazole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6478-73-5
:
Formula:C₇H₄Cl₂N₂
MDL Number:MFCD00211325
:187.03 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:208.0 to 212.0 deg-C
Specification Test Results
Frequently Bought Together