Order Entry
Italy
ContactUsLinkComponent
Dextran sulphate sodium salt, M.W. ~ 40000 (from dextran)
Dextran sulphate sodium salt, M.W. ~ 40000 (from dextran)
Catalog # TCIAD5144-25G
Supplier:  TCI
CAS Number:  
Dextran sulphate sodium salt, M.W. ~ 40000 (from dextran)
Catalog # TCIAD5144-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Sulfur:
    15.0 to 20.0 % (calcd.on dried substance)
  • Specific rotation [a]20/D:
    +75 to +105 deg (C=6, H2O,calcd.on dried substance)
  • limiting viscosity:
    0.05 to 0.13
  • Drying loss:
    max. 10.0 %

Specification Test Results

About this item

 :
Merck Index:14,02951
CAS Number:9011-18-1
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551