
2,3-Difluorobenzylamine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:72235-51-9
:
Formula:C₇H₇F₂N
MDL Number:MFCD00070793
:143.14 g/mol
Flash Pt:77°C
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together