Order Entry
Italy
ContactUsLinkComponent
 
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine ≥98.0% (by HPLC)
 :  TCI
 
undefined
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %

 

Frequently Bought Together