Order Entry
Italy
ContactUsLinkComponent
(S)-(-)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥97.0% (by GC)
 
(S)-(-)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥97.0% (by GC)
  TCIAD5107-1G
 :  TCI
 :  
undefined
 
(S)-(-)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥97.0% (by GC)
  TCIAD5107-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Optical purity (LC):
    min. 98.0 ee%
  • Melting point:
    142.0 to 146.0 deg-C

 

 

 :
 :452.1800 g/mol
 :GHS07
 :765278-73-7
 :C₂₀H₂₀Br₂O₂
 :MFCD03093977