Order Entry
Italy
ContactUsLinkComponent
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
Irritante
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI
Certificati
undefined
Irritante
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI

Risultati dei test sulle specifiche

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    81.0 to 85.0 deg-C

Risultati dei test sulle specifiche

Frequently Bought Together