
3,5-Dicarboxyphenylboronic acid (contains varying amounts of Anhydride)
TCIAD5073-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 109.0 %
:
:209.95 g/mol
:GHS07
:881302-73-4
:C₈H₇BO₆
:MFCD06203347