

1,5-Dimethyl-3-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
: TCI
:79080-31-2
:
:C₆H₇F₃N₂
:MFCD04972844
:164.13 g/mol
:71°C
:1993
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together