Order Entry
Italy
ContactUsLinkComponent
 
1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one ≥98.0% (by GC)
 :  TCI
 
undefined
1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    133.0 to 137.0 deg-C

 

Frequently Bought Together